missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ HC 030031

Description
Selective TRPA1 channel blocker that antagonizes AITC- and formalin-evoked calcium influx (IC50 values are 6.2 and 5.3 μM respectively). Does not block currents mediated by TRPV1, TRPV3, TRPV4, hERG or NaV1.2 channels. Inhibits AITC- and formalin-induced flinching in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 355.39 |
| Purity | >99% |
| Molecular Weight (g/mol) | 355.398 |
| Chemical Name or Material | HC 030031 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?