CAS RN 3371-27-5
CAS RN 3371-27-5
IUPAC Name:
2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol
Synonyms:
epigallocatechin
Molecular Weight (g/mol):
306.27
Molecular Formula:
C15H14O7
InChi Key:
XMOCLSLCDHWDHP-UHFFFAOYNA-N
SMILES:
OC1CC2=C(O)C=C(O)C=C2OC1C1=CC(O)=C(O)C(O)=C1