missing translation for 'onlineSavingsMsg'
Learn More

(-)-Gallocatechin

Product Code. 15403449
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15403449 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15403449 Supplier Thermo Scientific Alfa Aesar Supplier No. J60731.MA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3371-27-5 | C15H14O7 | 306.27 g/mol

Polyphenolic compound from green tea. Possesses free-radical scavenging ability

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Polyphenolic compound from green tea
  • Possesses free-radical scavenging ability
TRUSTED_SUSTAINABILITY

Specifications

CAS 3371-27-5
Color White
Molecular Formula C15H14O7
MDL Number MFCD01632616
Quantity 10 mg
Synonym --gallocatechin, 2s,3r-2-3,4,5-trihydroxyphenyl chroman-3,5,7-triol, 2s,3r-gallocatechin, ent-gallocatechin, 2s,3r-flavan-3,3',4',5,5',7-hexol, 2s,3r-flavan-3,5,7,3',4',5'-hexol, 2s,3r-2-3,4,5-trihydroxyphenyl-3,4-dihydro-2h-chromene-3,5,7-triol, 2s,3r---2-3,4,5-trihydroxyphenyl-3,4-dihydro-2h-chromene-3,5,7-triol, gallocatechingc, l-gallocatechin
InChI Key XMOCLSLCDHWDHP-DOMZBBRYSA-N
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)O
IUPAC Name (2S,3R)-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
Molecular Weight (g/mol) 306.27
PubChem CID 9882981
ChEBI CHEBI:71225
Formula Weight 306.27
Physical Form Powder
Chemical Name or Material (-)-Gallocatechin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.