accessibility menu, dialog, popup

UserName

CH3N(R)OH3C(R)HO(S)OH3C(S)(R)(R)OH(S)OCH3(S)OH(R)OCH3O(R)(R)OH3C(S)(R)(R)OO

CAS RN 302-27-2

IUPAC Name: (1S,2R,3R,4R,5R,6S,7S,8R,9R,13R,14R,16S,17S,18R)-8-(acetyloxy)-11-ethyl-5,7,14-trihydroxy-6,16,18-trimethoxy-13-(methoxymethyl)-11-azahexacyclo[7.7.2.1²,⁵.0¹,¹⁰.0³,⁸.0¹³,¹⁷]nonadecan-4-yl benzoate
Synonyms: aconitine
Molecular Weight (g/mol): 645.75
Molecular Formula: C34H47NO11
InChi Key: XFSBVAOIAHNAPC-XTHSEXKGSA-N
SMILES: CCN1C[C@@]2(COC)[C@H]3[C@@H](OC)[C@H]4C1[C@@]3([C@@H]1C[C@@]3(O)[C@H](OC(=O)C5=CC=CC=C5)[C@@H]1[C@]4(OC(C)=O)[C@@H](O)[C@@H]3OC)[C@H](C[C@H]2O)OC
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (2)
Concentration 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results