missing translation for 'onlineSavingsMsg'
Learn More

Aconitine, 98%

Product Code. 15424519
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15424519 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15424519 Supplier Thermo Scientific Alfa Aesar Supplier No. J62056.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 302-27-2 | C34H47NO11 | 645.746 g/mol

Opens tetrodotoxin-sensitive sodium channels. Useful for creating models of cardiac arrhythmia

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Opens tetrodotoxin-sensitive sodium channels
  • Useful for creating models of cardiac arrhythmia
TRUSTED_SUSTAINABILITY

Specifications

CAS 302-27-2
Melting Point 194°C to 202°C
Color White
Molecular Formula C34H47NO11
MDL Number MFCD00082375
Quantity 250 mg
UN Number UN1544
Beilstein 74608
Merck Index 14,118
Synonym aconitine
Solubility Information Soluble in ethanol; Sparingly soluble in water
InChI Key XFSBVAOIAHNAPC-OZUFZMTGSA-N
SMILES CCN1CC2(C(CC(C34C2C(C(C31)C5(C6C4CC(C6OC(=O)C7=CC=CC=C7)(C(C5O)OC)O)OC(=O)C)OC)OC)O)COC
Molecular Weight (g/mol) 645.746
PubChem CID 132470875
Formula Weight 645.75
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Aconitine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.