missing translation for 'onlineSavingsMsg'
Learn More

Aconitine, TRC

Product Code. 18073030
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18073030 10 mg 10mg
18070050 5 mg 5mg
18003630 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18073030 Supplier Toronto Research Chemicals Supplier No. TRCA18987510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-27-2
Molecular Formula C34 H47 N O11
Synonym Aconitane-3,8,13,14,15-pentol, 20-ethyl-1,6,16-trimethoxy-4-(methoxymethyl)-, 8-acetate 14-benzoate, (1α,3α,6α,14α,15α,16β)-∣2H-12,3,6a-Ethanylylidene-7,9-methanonaphth[2,3-b]azocine-4,8,9,11,11a(1H,7H)-pentol, 1-ethyldecahydro-6,10,13-trimethoxy-3-(methoxymethyl)-, 11a-acetate 8-benzoate (7CI)∣Aconitine (8CI)∣2H-12,3,6a-Ethanylylidene-7,9-methanonaphth[2,3-b]azocine, aconitane-3,8,13,14,15-pentol deriv.∣(1α,3α,6α,14α,15α,16β)-20-Ethyl-1,6,16-trimethoxy-4-(methoxymethyl)aconitane-3,8,13,14,15-pentol 8-acetate 14-benzoate

Specifications

Quantity 10 mg
Shipping Condition Room Temperature
Formula Weight 645.3149
Purity >95% (HPLC)
Molecular Weight (g/mol) 645.74, 645.75
InChI Key XFSBVAOIAHNAPC-XTHSEXKGSA-N
SMILES CCN1C[C@@]2(COC)[C@H]3[C@@H](OC)[C@H]4C1[C@@]3([C@@H]1C[C@@]3(O)[C@H](OC(=O)C5=CC=CC=C5)[C@@H]1[C@]4(OC(C)=O)[C@@H](O)[C@@H]3OC)[C@H](C[C@H]2O)OC
IUPAC Name (1S,2R,3R,4R,5R,6S,7S,8R,9R,13R,14R,16S,17S,18R)-8-(acetyloxy)-11-ethyl-5,7,14-trihydroxy-6,16,18-trimethoxy-13-(methoxymethyl)-11-azahexacyclo[7.7.2.1²,⁵.0¹,¹⁰.0³,⁸.0¹³,¹⁷]nonadecan-4-yl benzoate
Chemical Name or Material Aconitine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.