CAS RN 274901-16-5
CAS RN 274901-16-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Novus Biologicals™ Vildagliptin anti-diabetic agent
Vildagliptin is a potent inhibitor of dipeptidyl peptidase IV (DPP IV), an enzyme that cleaves glucagon-like peptide 1 (GLP-1) (1).
2
Vildagliptin, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Vildagliptin, MedChemExpress
MedChemExpress Vildagliptin (LAF237) is a potent, stable, selective dipeptidyl peptidase IV (DPP-IV) inhibitor with an IC50 of 3.5 nM in human Caco-2 cells. Vildagliptin possesses excellent oral bioavailability and potent antihyperglycemic activity.