missing translation for 'onlineSavingsMsg'
Learn More

Vildagliptin, MedChemExpress

Product Code. 16466436
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16466436

Brand: Medchem Express HY14291

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vildagliptin (LAF237) is a potent, stable, selective dipeptidyl peptidase IV (DPP-IV) inhibitor with an IC50 of 3.5 nM in human Caco-2 cells. Vildagliptin possesses excellent oral bioavailability and potent antihyperglycemic activity.

Vildagliptin (LAF237) is a potent, stable, selective dipeptidyl peptidase IV (DPP-IV) inhibitor with an IC50 of 3.5 nM in human Caco-2 cells. Vildagliptin possesses excellent oral bioavailability and potent antihyperglycemic activity.

Specifications

Chemical Name or Material Vildagliptin
CAS 274901-16-5
Color White
Purity Grade Notes Research
Molecular Formula C17H25N3O2
Quantity 100 mg
Synonym LAF237 NVP-LAF 237
Solubility Information H2O : 50 mg/mL (164.80 mM; Need ultrasonic)
SMILES O[C@@]1(C2)C[C@H]3C[C@H](C[C@]2(NCC(N4[C@H](C#N)CCC4)=O)C3)C1
Molecular Weight (g/mol) 303.4
Formula Weight 303.4
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.