CAS RN 23256-42-0
IUPAC Name:
2-hydroxypropanoic acid; 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
Synonyms:
lactic acid; trimethoprim
Molecular Weight (g/mol):
380.4
Molecular Formula:
C17H24N4O6
InChi Key:
IIZVTUWSIKTFKO-UHFFFAOYNA-N
SMILES:
CC(O)C(O)=O.COC1=CC(CC2=CN=C(N)N=C2N)=CC(OC)=C1OC
1
–
3
of
3
results
2
Melford Trimethoprim Lactate
Trimethoprim lactate is a combination of the antibiotic trimethoprim and organic compound lactate.
3
Trimethoprim Lactate Salt, TRC
CAS: 23256-42-0 Molecular Formula: C14H18N4O3 . C3H6O3 Molecular Weight (g/mol): 380.4 Synonym: 5-[(3,4,5-Trimethoxyphenyl)methyl]-2,4-pyrimidinediamine Mono(2-hydroxypropanoate),Lactotrim,Trimethoprim Lactate; IUPAC Name: 2-hydroxypropanoic acid;5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine SMILES: COc1cc(Cc2cnc(N)nc2N)cc(OC)c1OC.CC(O)C(=O)O