missing translation for 'onlineSavingsMsg'
Learn More

Trimethoprim lactate, 98%

Product Code. 15458089
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Plastic Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15458089 250 mg Plastic Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15458089 Supplier Thermo Scientific Alfa Aesar Supplier No. J66646.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trimethoprim, CAS # 738-70-5, is a trimethoxybenzyl-pyrimidine used as an antimicrobial. | CAS: 23256-42-0 | C17H24N4O6 | 380.401 g/mol

A broad spectrum antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Trimethoprim is a synthetic pyrimidine analog that binds dihydrofolate-reductase and inhibits the synthesis of folic acid; when paired with sulfamethoxazole, its activity is enhanced

• Trimethoprim has antibacterial and antiprotozoal properties

Applications

• Trimethoprim inhibits the conversion of para-aminobenzoic acid (PABA) to folic acid near the end of the synthesis pathway and can be used to study related effects

• Trimethoprim has been radiolabeled and used to trace routes of infection, and it also has been used experimentally as a fluorescent labeler to quantify dihydrofolate-reductase expression

TRUSTED_SUSTAINABILITY

Specifications

Type Trimethoprim Lactate
Physical Form Powder
CAS 23256-42-0
Packaging Plastic Bottle
Quantity 250 mg
Molecular Formula C17H24N4O6
MDL Number MFCD00171722
Synonym 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine lactate salt
Solubility Information Soluble in water
InChI Key IIZVTUWSIKTFKO-UHFFFAOYSA-N
SMILES CC(C(=O)O)O.COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
IUPAC Name 2-hydroxypropanoic acid;5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
Molecular Weight (g/mol) 380.401
PubChem CID 3084396
Formula Weight 380.39
Percent Purity 98%
Chemical Name or Material Trimethoprim lactate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.