missing translation for 'onlineSavingsMsg'
Learn More

Trimethoprim Lactate Salt, TRC

Product Code. 30179946
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179946 250 mg 250mg
30179803 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179946 Supplier Toronto Research Chemicals Supplier No. TRCT795628250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23256-42-0
Molecular Formula C14H18N4O3 . C3H6O3
Molecular Weight (g/mol) 380.4
Synonym 5-[(3,4,5-Trimethoxyphenyl)methyl]-2,4-pyrimidinediamine Mono(2-hydroxypropanoate), Lactotrim, Trimethoprim Lactate;
IUPAC Name 2-hydroxypropanoic acid;5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES COc1cc(Cc2cnc(N)nc2N)cc(OC)c1OC.CC(O)C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H18N4O3.C3H6O3/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15;1-2(4)3(5)6/h5-7H,4H2,1-3H3,(H4,15,16,17,18);2,4H,1H3,(H,5,6)
Formula Weight 380.1696
Chemical Name or Material Trimethoprim Lactate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.