missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Trimethoprim Lactate
Description
Trimethoprim lactate is a combination of the antibiotic trimethoprim and organic compound lactate.
Trimethoprim is a bacteriostatic antibiotic that inhibits bacterial dihydrofolate reductase, an enzyme involved in the synthesis of tetrahydrofolic acid. This acid is crucial for the production of nucleic acids (DNA and RNA) in bacteria. By blocking this enzyme, trimethoprim disrupts the folate synthesis pathway, leading to the inhibition of bacterial growth. Trimethoprim is frequently used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its presence in the culture medium allows for the specific isolation of bacteria that lack resistance to trimethoprim.
Specifications
Specifications
| Chemical Name or Material | Trimethoprim Lactate |
| CAS | 23256-42-0 |
| Quantity | 250 mg |
| Molecular Formula | C14H18N4O3-C3H6O3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?