CAS RN 220904-83-6
CAS RN 220904-83-6
IUPAC Name:
3-[(3,5-dibromo-4-hydroxyphenyl)methylidene]-5-iodo-2,3-dihydro-1H-indol-2-one
Synonyms:
3-[(3,5-dibromo-4-hydroxyphenyl)methylidene]-5-iodo-1H-indol-2-one
Molecular Weight (g/mol):
520.95
Molecular Formula:
C15H8Br2INO2
InChi Key:
LMXYVLFTZRPNRV-UHFFFAOYSA-N
SMILES:
OC1=C(Br)C=C(C=C2C(=O)NC3=C2C=C(I)C=C3)C=C1Br
1
–
2
of
2
results
1
Tocris Bioscience™ GW 5074
CAS: 220904-83-6 Molecular Formula: C15H8Br2INO2
2
GW 5074, MedChemExpress
MedChemExpress GW 5074 is a potent and selective c-Raf inhibitor with IC50 of 9 nM, and has no effect on the activities of JNK1/2/3, MEK1, MKK6/7, CDK1/2, c-Src, p38 MAP, VEGFR2 or c-Fms.