missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GW 5074
GREENER_CHOICE

Product Code. 18075962
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18075962

Brand: Tocris Bioscience™ 1381/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective c-Raf1 kinase inhibitor

GW 5074 is a potent, selective and cell-permeable c-Raf1 kinase inhibitor (IC50 = 9 nM). GW 5074 displays ≥ 100-fold selectivity for raf kinase over CDK1, CDK2, c-src, ERK2, MEK, p38, Tie2, VEGFR2 and c-fm. GW 5074 binds mutant huntingtin (mHTT) and decreases levels of mHTT in cultured hippocampal neurons and fibroblasts from patients with Huntington's disease. Brain penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220904-83-6
Molecular Formula C15H8Br2INO2

Specifications

Quantity 50 mg
Formula Weight 520.94
Purity >98%
Molecular Weight (g/mol) 520.95
Chemical Name or Material GW 5074
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.