accessibility menu, dialog, popup

UserName

CAS RN 216144-68-2

FFFFClO

CAS RN 216144-68-2

IUPAC Name: 3-fluoro-4-(trifluoromethyl)benzoyl chloride
Synonyms: 3-fluoro-4-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 226.55
Molecular Formula: C8H3ClF4O
InChi Key: DPDAEMNQJYDHRQ-UHFFFAOYSA-N
SMILES: FC1=CC(=CC=C1C(F)(F)F)C(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results