missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is employed as an active pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture sensitive. Store away from alcohols, amines, oxidizing agents and active metals. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is employed as an active pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture sensitive. Store away from alcohols, amines, oxidizing agents and active metals. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Specifications
| Density | 1.486 |
| Boiling Point | 151°C to 153°C |
| Flash Point | 81°C (177°F) |
| Refractive Index | 1.4656 |
| Quantity | 5 g |
| UN Number | UN3265 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Not miscible or difficult to mix in water. |
| Formula Weight | 226.56 |
| Percent Purity | 97% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction