CAS RN 215030-90-3
CAS RN 215030-90-3
IUPAC Name:
4-{2-[5,5-dimethyl-8-(2-phenylethynyl)-5,6-dihydronaphthalen-2-yl]ethenyl}benzoic acid
Synonyms:
4-{2-[5,5-dimethyl-8-(2-phenylethynyl)-6H-naphthalen-2-yl]ethenyl}benzoic acid
Molecular Weight (g/mol):
404.51
Molecular Formula:
C29H24O2
InChi Key:
YCADIXLLWMXYKW-UHFFFAOYSA-N
SMILES:
CC1(C)CC=C(C#CC2=CC=CC=C2)C2=C1C=CC(C=CC1=CC=C(C=C1)C(O)=O)=C2