missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies BMS 493

Product Code. 17150302
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17150302

Brand: STEMCELL Technologies 73974

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Retinoid pathway activator; Activates pan-retinoic acid receptor

  • BMS 493 is an inverse agonist of the pan-retinoic acid receptors (RARs)
  • RARs are ligand-dependent transcription factors that regulate gene networks that control cell growth, differentiation, survival, and death (Germain et al.) BMS 493 has been shown to increase nuclear receptor co-repressor (NCoR) interaction with RARs (Germain et al.; Rinkevich et al.)
  • Affects development of bronchial tubule formation in mice (Chazaud et al.)
  • Inhibits activation of immature dendritic cells (Geissmann et al.)
  • Inhibits differentiation of leukemic blasts (Kamashev et al.)

Specifications

CAS 215030-90-3
Synonym BMS 204493
Quantity 10 mg
Molecular Formula C29H24O2
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.