missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BMS 493
GREENER_CHOICE

Product Code. 18095692
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18095692

Brand: Tocris Bioscience™ 3509/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pan-RAR inverse agonist

Pan-retinoic acid receptor (pan-RAR) inverse agonist. Enhances nuclear corepressor (NCoR) interaction with RARs. Binding induces analogous conformational changes in all RAR types (RARα, RARβ and RARγ).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 215030-90-3
Molecular Formula C29H24O2

Specifications

Quantity 50 mg
Formula Weight 404.5
Purity >98%
Molecular Weight (g/mol) 404.509
Chemical Name or Material BMS 493
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.