accessibility menu, dialog, popup

UserName

CAS RN 2096335-33-8

BONH3CFHOOH

CAS RN 2096335-33-8

IUPAC Name: [2-fluoro-5-(4-methylpiperidine-1-carbonyl)phenyl]boronic acid
Synonyms: 2-fluoro-5-(4-methylpiperidine-1-carbonyl)phenylboronic acid
Molecular Weight (g/mol): 265.09
Molecular Formula: C13H17BFNO3
InChi Key: PBHCGBUAOXBGIF-UHFFFAOYSA-N
SMILES: CC1CCN(CC1)C(=O)C1=CC(B(O)O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results