missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(4-methyl-1-piperidinylcarbonyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15421416
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15421416 250 mg 250mg
15411416 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15421416

Brand: Thermo Scientific Alfa Aesar H54626.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2096335-33-8
Molecular Formula C13H17BFNO3
Molecular Weight (g/mol) 265.09
MDL Number MFCD20265302
InChI Key PBHCGBUAOXBGIF-UHFFFAOYSA-N
Synonym 2-fluoro-5-4-methyl-1-piperidinylcarbonyl phenylboronic acid, 2-fluoro-5-4-methyl-1-piperidinylcarbonyl benzeneboronic acid
PubChem CID 73995899

Specifications

Quantity 250 mg
Formula Weight 265.09
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(4-methyl-1-piperidinylcarbonyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.