accessibility menu, dialog, popup

UserName

CAS RN 205748-03-4

H3COONOO

CAS RN 205748-03-4

IUPAC Name: 1-{4-[(4-nitrophenyl)methoxy]phenyl}ethan-1-one
Synonyms: 1-{4-[(4-nitrophenyl)methoxy]phenyl}ethanone
Molecular Weight (g/mol): 271.27
Molecular Formula: C15H13NO4
InChi Key: GUBOWULDVFKCRF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(OCC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results