missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Nitrobenzyloxy)acetophenone, 97%, Thermo Scientific™

Product Code. 11492748
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11492748

Brand: Thermo Scientific Alfa Aesar A16738.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205748-03-4
Molecular Formula C15H13NO4
Molecular Weight (g/mol) 271.27
MDL Number MFCD00075690
InChI Key GUBOWULDVFKCRF-UHFFFAOYSA-N
Synonym 4'-4-nitrobenzyloxy acetophenone, 1-4-4-nitrobenzyl oxy phenyl ethanone, 4-4-nitrobenzyloxy acetophenone, 1-4-4-nitrophenyl methoxy phenyl ethanone, maybridge1_001947, acmc-1chn1, 1-acetyl-4-4-nitrophenyl methoxy benzene
PubChem CID 2801340
SMILES CC(=O)C1=CC=C(OCC2=CC=C(C=C2)[N+]([O-])=O)C=C1

Specifications

Melting Point 129°C to 131°C
Quantity 25 g
Beilstein 8321694
IUPAC Name 1-{4-[(4-nitrophenyl)methoxy]phenyl}ethan-1-one
Formula Weight 271.28
Percent Purity 97%
Chemical Name or Material 4'-(4-Nitrobenzyloxy)acetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.