accessibility menu, dialog, popup

UserName

CAS RN 205652-55-7

SSSSSSOOOOOOOOOOOOHOOHHOOHOHOHOHHOHOOHOHOH

CAS RN 205652-55-7

IUPAC Name: 37,38,39,40,41,42-hexahydroxyheptacyclo[31.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹]dotetraconta-1(37),3,5,7(42),9,11,13(41),15(40),16,18,21(39),22,24,27(38),28,30,33,35-octadecaene-5,11,17,23,29,35-hexasulfonic acid hydrate
Synonyms: 37,38,39,40,41,42-hexahydroxyheptacyclo[31.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹]dotetraconta-1(37),3,5,7(42),9,11,13(41),15(40),16,18,21(39),22,24,27(38),28,30,33,35-octadecaene-5,11,17,23,29,35-hexasulfonic acid hydrate
Molecular Weight (g/mol): 1135.1
Molecular Formula: C42H38O25S6
InChi Key: SWWAZWAJPNREHZ-UHFFFAOYSA-N
SMILES: O.OC1=C2CC3=CC(=CC(CC4=CC(=CC(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(CC1=CC(=C2)S(O)(=O)=O)=CC(=C7)S(O)(=O)=O)=CC(=C6)S(O)(=O)=O)=CC(=C5)S(O)(=O)=O)=C4O)S(O)(=O)=O)=C3O)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results