missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfonic-calix[6]arene hydrate, Alfa Aesar™

Product Code. 11353177
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11353177

Brand: Thermo Scientific Alfa Aesar L13625.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 205652-55-7
Molecular Formula C42H36O24S6
Molecular Weight (g/mol) 1117.09
MDL Number MFCD00216907,MFCD00143376
InChI Key BXOUPTJVBGEDIR-UHFFFAOYSA-N
Synonym 4-sulfocalix 6 arene, 4-sulfocalix 6 arene hydrate, hexahydroxycalix 6 arenehexasulfonic acid, 4-sulfonic calix 6 arene, acmc-20m53e, calix 6 arene-p-sulfonate, hexasulfo hexahydroxy calix 6 arene
PubChem CID 2724887
IUPAC Name 37,38,39,40,41,42-hexahydroxyheptacyclo[31.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹]dotetraconta-1(37),3,5,7(42),9,11,13(41),15(40),16,18,21(39),22,24,27(38),28,30,33,35-octadecaene-5,11,17,23,29,35-hexasulfonic acid
SMILES OC1=C2CC3=CC(=CC(CC4=CC(=CC(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(CC1=CC(=C2)S(O)(=O)=O)=CC(=C7)S(O)(=O)=O)=CC(=C6)S(O)(=O)=O)=CC(=C5)S(O)(=O)=O)=C4O)S(O)(=O)=O)=C3O)S(O)(=O)=O

Specifications

Melting Point ∽246°C (decomposition)
Beilstein 5231405
Solubility Information Soluble in water.
Quantity 5 g
Formula Weight 1117.12 (Anhydrous)
Chemical Name or Material 4-Sulfonic-calix[6]arene hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.