CAS RN 2009-24-7
CAS RN 2009-24-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Xanthotoxol, TRC
CAS: 2009-24-7 Molecular Formula: C11 H6 O4 Molecular Weight (g/mol): 202.16 Synonym: 7H-Furo[3,2-g][1]benzopyran-7-one, 9-hydroxy-,5-Benzofuranacrylic acid, 6,7-dihydroxy-, δ-lactone (7CI),Xanthotoxol (6CI),2-Propenoic acid, 3-(6,7-dihydroxy-5-benzofuranyl)-, δ-lactone,8-Hydroxypsoralen,9-Hydroxy-7H-furo[3,2-g][1]benzopyran-7-one,NSC 401269,Psoralen, 8-hydroxy- IUPAC Name: 9-hydroxyfuro[3,2-g]chromen-7-one SMILES: Oc1c2OC(=O)C=Cc2cc3ccoc13
2
Xanthotoxol, MedChemExpress
MedChemExpress Xanthotoxol (8-Hydroxypsoralen) is a biologically active linear furocoumarin, shows strong pharmacological activities as anti-inflammatory, antioxidant, 5-HT antagonistic, and neuroprotective effects.