missing translation for 'onlineSavingsMsg'
Learn More

Xanthotoxol, TRC

Product Code. 30455293
Change view
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
1g
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455293 250 mg 250mg
30454060 1 g 1g
30453484 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455293 Supplier Toronto Research Chemicals Supplier No. TRCX742500250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2009-24-7
Molecular Formula C11 H6 O4
Molecular Weight (g/mol) 202.16
Synonym 7H-Furo[3,2-g][1]benzopyran-7-one, 9-hydroxy-, 5-Benzofuranacrylic acid, 6,7-dihydroxy-, δ-lactone (7CI), Xanthotoxol (6CI), 2-Propenoic acid, 3-(6,7-dihydroxy-5-benzofuranyl)-, δ-lactone, 8-Hydroxypsoralen, 9-Hydroxy-7H-furo[3,2-g][1]benzopyran-7-one, NSC 401269, Psoralen, 8-hydroxy-
IUPAC Name 9-hydroxyfuro[3,2-g]chromen-7-one
SMILES Oc1c2OC(=O)C=Cc2cc3ccoc13

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H6O4/c12-8-2-1-6-5-7-3-4-14-10(7)9(13)11(6)15-8/h1-5,13H
Formula Weight 202.0266
Chemical Name or Material Xanthotoxol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.