missing translation for 'onlineSavingsMsg'
Learn More

Xanthotoxol, MedChemExpress

Product Code. 16487796
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487796 100 mg 100mg
16473947 50 mg 50mg
16428028 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16487796 Supplier Medchem Express Supplier No. HY30152
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Xanthotoxol (8-Hydroxypsoralen) is a biologically active linear furocoumarin, shows strong pharmacological activities as anti-inflammatory, antioxidant, 5-HT antagonistic, and neuroprotective effects.

Xanthotoxol (8-Hydroxypsoralen) is a biologically active linear furocoumarin, shows strong pharmacological activities as anti-inflammatory, antioxidant, 5-HT antagonistic, and neuroprotective effects.

Specifications

Chemical Name or Material Xanthotoxol
CAS 2009-24-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H6O4
Quantity 100 mg
Synonym 8-Hydroxypsoralen
Solubility Information DMSO : 100 mg/mL (494.66 mM; Need ultrasonic)
SMILES O=C1OC(C(C=C1)=C2)=C(O)C3=C2C=CO3
Molecular Weight (g/mol) 202.16
Formula Weight 202.16
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.