CAS RN 1956356-56-1
CAS RN 1956356-56-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
KYA1797K, MedChemExpress
MedChemExpress KYA1797K is a potent and selective Wnt/β-catenin inhibitor with an IC50 of 0.75 μM.
2
Tocris Bioscience™ KYA 1797K
Wnt/β-catenin signaling inhibitor