missing translation for 'onlineSavingsMsg'
Learn More

KYA1797K, MedChemExpress

Product Code. 16483258
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483258

Brand: Medchem Express HY101090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KYA1797K is a potent and selective Wnt/β-catenin inhibitor with an IC50 of 0.75 μM.

KYA1797K is a potent and selective Wnt/β-catenin inhibitor with an IC50 of 0.75 μM.

Specifications

Chemical Name or Material KYA1797K
CAS 1956356-56-1
Color Orange
Purity Grade Notes Research
Molecular Formula C17H11KN2O6S2
Quantity 10 mg
Solubility Information DMSO : 6 mg/mL (13.56 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.26 mM; Need ultrasonic)
SMILES O=C(O[K])CCN(C/1=O)C(SC1=C/C2=CC=C(C3=CC=C([N+]([O-])=O)C=C3)O2)=S
Molecular Weight (g/mol) 442.51
Formula Weight 442.51
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.