missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KYA 1797K
GREENER_CHOICE

Product Code. 18799356
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18799356

Brand: Tocris Bioscience™ 7002/25

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Wnt/β-catenin signaling inhibitor

KYA 1797K is a Wnt/β-catenin signaling inhibitor (IC50 = 0.75 μM). Increases binding affinity of β-catenin for axin, GSK-3β and β-TrCP. Promotes formation of the destruction complex, activation of GSK-3β and phosphorylation of β-catenin and K-Ras, leading to their degradation via the ubiquitin/proteasome system. Reduces levels of β-catenin and K-Ras and inhibits growth of colorectal cancer (CRC) cells in vitro and in mouse xenograft models.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5Z)-5-[[5-(4-Nitrophenyl)-2-furanyl]methylene]-4-oxo-2-thioxo-3-thiazolidinepropanoic acid potassium salt
CAS 1956356-56-1
Quantity 25 mg
Molecular Formula C17H11KN2O6S2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.