accessibility menu, dialog, popup

UserName

CAS RN 1530-34-3

BrH3CCH3P

CAS RN 1530-34-3

IUPAC Name: (3-methylbut-2-en-1-yl)triphenylphosphanium bromide
Synonyms: (3-methylbut-2-en-1-yl)triphenylphosphanium bromide
Molecular Weight (g/mol): 411.32
Molecular Formula: C23H24BrP
InChi Key: FTMCNCGWNJMMQS-UHFFFAOYSA-M
SMILES: [Br-].CC(C)=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results