missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Methyl-2-butenyl)triphenyl-phosphonium Bromide, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (3-Methyl-2-butenyl)triphenyl-phosphonium Bromide |
| CAS | 1530-34-3 |
| Molecular Formula | C23H24BrP |
| Quantity | 1 g |
| Synonym | (3,3-Dimethylallyl)triphenylphosphonium Bromide, (3-Methylbut-2-enyl)triphenylphosphonium Bromide, NSC 219242 |
| InChI Formula | InChI=1S/C23H24P.BrH/c1-20(2)18-19-24(21-12-6-3-7-13-21,22-14-8-4-9-15-22)23-16-10-5-11-17-23;/h3-18H,19H2,1-2H3;1H/q+1;/p-1 |
| SMILES | [Br-].CC(=CC[P+](c1ccccc1)(c2ccccc2)c3ccccc3)C |
| Molecular Weight (g/mol) | 411.31 |
| Formula Weight | 410.0799 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction