missing translation for 'onlineSavingsMsg'
Learn More

(3-Methyl-2-butenyl)triphenyl-phosphonium Bromide, TRC

Product Code. 30470486
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
This item is not returnable. View return policy

Product Code. 30470486

Brand: Toronto Research Chemicals TRCM294345500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3-Methyl-2-butenyl)triphenyl-phosphonium Bromide
CAS 1530-34-3
Molecular Formula C23H24BrP
Quantity 500 mg
Synonym (3,3-Dimethylallyl)triphenylphosphonium Bromide, (3-Methylbut-2-enyl)triphenylphosphonium Bromide, NSC 219242
InChI Formula InChI=1S/C23H24P.BrH/c1-20(2)18-19-24(21-12-6-3-7-13-21,22-14-8-4-9-15-22)23-16-10-5-11-17-23;/h3-18H,19H2,1-2H3;1H/q+1;/p-1
SMILES [Br-].CC(=CC[P+](c1ccccc1)(c2ccccc2)c3ccccc3)C
Molecular Weight (g/mol) 411.31
Formula Weight 410.0799
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.