CAS RN 150566-71-5
CAS RN 150566-71-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Levamlodipine besylate, MedChemExpress
MedChemExpress Levamlodipine besylate ((S)-Amlodipine besylate) is a powerful dihydropyridine calcium channel blocker, possessing vasodilation properties and used in the treatment of hypertension and angina.