missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levamlodipine besylate, MedChemExpress
Description
Levamlodipine besylate ((S)-Amlodipine besylate) is a powerful dihydropyridine calcium channel blocker, possessing vasodilation properties and used in the treatment of hypertension and angina.
Specifications
Specifications
| Chemical Name or Material | Levamlodipine besylate |
| CAS | 150566-71-5 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C26H31ClN2O8S |
| Quantity | 50 mg |
| Synonym | (S)-Amlodipine besylate Levoamlodipine besylate |
| Solubility Information | DMSO : ≥ 150 mg/mL (264.53 mM) |
| SMILES | O=C(C1=C(COCCN)NC(C)=C(C(OC)=O)[C@@H]1C2=CC=CC=C2Cl)OCC.OS(C3=CC=CC=C3)(=O)=O |
| Molecular Weight (g/mol) | 567.05 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?