CAS RN 1378872-36-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
ML239, MedChemExpress
MedChemExpress ML239 is a potent and selective inhibitor of breast cancer stem cells, with an IC50 of 1.16 μM.
2
ML 239
Breast cancer stem cell inhibitor