missing translation for 'onlineSavingsMsg'
Learn More

ML 239
GREENER_CHOICE

Product Code. 18796886
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18796886 25 mg 25mg
18707576 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18796886 Supplier Tocris Bioscience Supplier No. 4829/25

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Breast cancer stem cell inhibitor

ML 239 is a breast cancer stem cell inhibitor (IC50 = 1.16 μM). Exhibits 24-fold selectivity for breast cancer stem cells over normal mammary epithelial cells. Also cytotoxic towards MDA-MB-231 breast cancer cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2,4,6-Trichlorophenoxy)acetic acid (2E)-2-(1H-pyrrol-2-ylmethylene)hydrazide
CAS 1378872-36-6
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C13H10Cl3N3O2
Formula Weight Observed MW: 346.6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.