missing translation for 'onlineSavingsMsg'
Learn More

ML239, MedChemExpress

Product Code. 16497988
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497988 10 mg 10mg
16463917 50 mg 50mg
16430458 5 mg 5mg
16407776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497988 Supplier Medchem Express Supplier No. HY19971

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML239 is a potent and selective inhibitor of breast cancer stem cells, with an IC50 of 1.16 μM.

ML239 is a potent and selective inhibitor of breast cancer stem cells, with an IC50 of 1.16 μM.

Specifications

Chemical Name or Material ML239
CAS 1378872-36-6
Color White
Purity Grade Notes Research
Molecular Formula C13H10Cl3N3O2
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (288.52 mM; Need ultrasonic)
SMILES O=C(N/N=C/C1=CC=CN1)COC2=C(Cl)C=C(Cl)C=C2Cl
Molecular Weight (g/mol) 346.6
Formula Weight 346.6
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.