CAS RN 1309793-47-2
IUPAC Name:
2-[3-(2,4-difluorophenyl)-4,5-dihydro-1,2-oxazol-5-yl]-1-(morpholin-4-yl)ethan-1-one
Synonyms:
2-[3-(2,4-difluorophenyl)-4,5-dihydro-1,2-oxazol-5-yl]-1-(morpholin-4-yl)ethanone
Molecular Weight (g/mol):
310.3
Molecular Formula:
C15H16F2N2O3
InChi Key:
GUWOSEVHCSNYFK-UHFFFAOYNA-N
SMILES:
FC1=CC(F)=C(C=C1)C1=NOC(CC(=O)N2CCOCC2)C1
1
–
2
of
2
results
1
(±)-CPSI-1306, MedChemExpress
MedChemExpress (±)-CPSI-1306 is an orally available antagonist of macrophage migration inhibitory factor (MIF).