missing translation for 'onlineSavingsMsg'
Learn More

(±)-CPSI-1306, MedChemExpress

Product Code. 16480677
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480677 50 mg 50mg
16445266 100 mg 100mg
16423046 25 mg 25mg
16414358 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16480677 Supplier Medchem Express Supplier No. HY110095

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (±)-CPSI-1306 is an orally available antagonist of macrophage migration inhibitory factor (MIF).

(±)-CPSI-1306 is an orally available antagonist of macrophage migration inhibitory factor (MIF).

Specifications

Chemical Name or Material (±)-CPSI-1306
CAS 1309793-47-2
Color White
Purity Grade Notes Research
Molecular Formula C15H16F2N2O3
Quantity 50 mg
Solubility Information DMSO : 6 mg/mL (19.34 mM; Need ultrasonic)
SMILES FC1=C(C=CC(F)=C1)C2=NOC(CC(N3CCOCC3)=O)C2
Molecular Weight (g/mol) 310.3
Formula Weight 310.3
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.