missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-CPSI 1306, Tocris Bioscience™

Description
(±)-CPSI 1306 is reported to be a macrophage inhibitory factor (MIF) inhibitor.
This product is racemic.
Specifications
Specifications
| CAS | 1309793-47-2 |
| Molecular Formula | C15H16F2N2O3 |
| Quantity | 10 mg |
| Synonym | inverted exclamation marka-cpsi 1306 |
| InChI Key | GUWOSEVHCSNYFK-NSHDSACASA-N |
| SMILES | C1COCCN1C(=O)CC2CC(=NO2)C3=C(C=C(C=C3)F)F |
| IUPAC Name | 2-[(5S)-3-(2,4-difluorophenyl)-4,5-dihydro-1,2-oxazol-5-yl]-1-morpholin-4-ylethanone |
| Molecular Weight (g/mol) | 310.301 |
| PubChem CID | 97992476 |
| Formula Weight | 310.3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?