CAS RN 130477-52-0
CAS RN 130477-52-0
IUPAC Name:
ethyl 15-methoxy-12-oxo-2,4,11-triazatetracyclo[11.4.0.0²,⁶.0⁷,¹¹]heptadeca-1(13),3,5,14,16-pentaene-5-carboxylate
Synonyms:
ethyl 15-methoxy-12-oxo-2,4,11-triazatetracyclo[11.4.0.0²,⁶.0⁷,¹¹]heptadeca-1(13),3,5,14,16-pentaene-5-carboxylate
Molecular Weight (g/mol):
341.37
Molecular Formula:
C18H19N3O4
InChi Key:
YKYOQIXTECBVBB-UHFFFAOYNA-N
SMILES:
CCOC(=O)C1=C2C3CCCN3C(=O)C3=C(C=CC(OC)=C3)N2C=N1
1
–
2
of
2
results
1
2
L-655708, MedChemExpress
MedChemExpress L-655708 is a potent α5 subunit-selective GABAA receptor inverse agonist (Ki=0.45 nM).