missing translation for 'onlineSavingsMsg'
Learn More

L-655708, MedChemExpress

Product Code. 16492197
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492197 50 mg 50mg
16468667 5 mg 5mg
16406068 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492197 Supplier Medchem Express Supplier No. HY14426

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-655708 is a potent α5 subunit-selective GABAA receptor inverse agonist (Ki=0.45 nM).

L-655708 is a potent α5 subunit-selective GABAA receptor inverse agonist (Ki=0.45 nM).

Specifications

Chemical Name or Material L-655708
CAS 130477-52-0
Color White
Purity Grade Notes Research
Molecular Formula C18H19N3O4
Quantity 50 mg
Solubility Information DMSO : 20 mg/mL (58.59 mM; Need ultrasonic)
SMILES COC1=CC=C(C2=C1)N3C=NC(C(OCC)=O)=C3[C@@]4([H])N(CCC4)C2=O
Molecular Weight (g/mol) 341.36
Formula Weight 341.36
Percent Purity 99.08%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.