accessibility menu, dialog, popup

UserName

CAS RN 126643-37-6

CH3OO(R)(R)O(S)H3CNNCH3ONOH3C

CAS RN 126643-37-6

IUPAC Name: methyl (15S,16R,18R)-16-methoxy-4,15-dimethyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate
Synonyms: methyl (15S,16R,18R)-16-methoxy-4,15-dimethyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate
Molecular Weight (g/mol): 495.54
Molecular Formula: C29H25N3O5
InChi Key: QTYMDECKVKSGSM-YMUMJAELSA-N
SMILES: COC(=O)[C@]1(C[C@H]2O[C@]1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CN(C)C1=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results