missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ KT 5823
Description
Selective inhibitor of protein kinase G (Ki values are 0.23, 4 and > 10 μM for inhibition of PKG, PKC and PKA respectively). Inhibits SNP-stimulated PKG activity with an IC50 of 60 nM in dispersed smooth muscle cells and has little effect on PKA activity at concentrations of up to 10 μM.
Specifications
Specifications
| CAS | 126643-37-6 |
| Quantity | 100 μg |
| Molecular Formula | C29H25N3O5 |
| Formula Weight | 495.53 |
| Purity | 95% |
| Molecular Weight (g/mol) | 495.535 |
| Chemical Name or Material | KT 5823 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?