CAS RN 125-10-0
CAS RN 125-10-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Prednisone Acetate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Prednisone Acetate, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
3
Prednisone acetate, MedChemExpress
MedChemExpress Prednisone acetate (Prednisone 21-acetate), the acetate salt form of prednisolone, is a glucocorticoid receptor agonist with anti-inflammatory and immunomodulating properties.