missing translation for 'onlineSavingsMsg'
Learn More

Prednisone Acetate, Mikromol™

Product Code. 17920766
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17920766

Brand: Mikromol MM0196.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Prednisone Acetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prednisone Acetate
CAS 125-10-0
Analyte or Component Names Prednisone Acetate
Shipping Condition Room Temperature
Molecular Formula C23 H28 O6
Certifications/Compliance ISO 17034
Synonym Prednisone Acetate, 21-(Acetyloxy)-17-hydroxypregna-1,4-diene-3,11,20-trione, 17,21-Dihydroxypregna-1,4-diene-3,11,20-trione 21-acetate, Cortancyl, Delcortin, Delta-Corlin, Deltalone, Ferrosan, NSC 10965, Nisone, Prednisone 21-acetate, Prednisone acetate, 21-Acetoxy-17alpha-hydroxypregna-1,4-diene-3,11,20-trione, Delta1-Cortisone 21-acetate, Delta1-Dehydrocortisone acetate, Prednisone Impurity E
InChI Formula InChI=1S/C23H28O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h6,8,10,16-17,20,28H,4-5,7,9,11-12H2,1-3H3/t16-,17-,20+,21-,22-,23-/m0/s1
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3C(=O)C[C@]12C
IUPAC Name [2-[(8S,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3,11-dioxo-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 400.46
Quantity 250 mg
Formula Weight 400.1886 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.