missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prednisone Acetate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Prednisone Acetate |
| CAS | 125-10-0 |
| Analyte or Component Names | Prednisone Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H28 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Prednisone Acetate, 21-(Acetyloxy)-17-hydroxypregna-1,4-diene-3,11,20-trione, 17,21-Dihydroxypregna-1,4-diene-3,11,20-trione 21-acetate, Cortancyl, Delcortin, Delta-Corlin, Deltalone, Ferrosan, NSC 10965, Nisone, Prednisone 21-acetate, Prednisone acetate, 21-Acetoxy-17alpha-hydroxypregna-1,4-diene-3,11,20-trione, Delta1-Cortisone 21-acetate, Delta1-Dehydrocortisone acetate, Prednisone Impurity E |
| InChI Formula | InChI=1S/C23H28O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h6,8,10,16-17,20,28H,4-5,7,9,11-12H2,1-3H3/t16-,17-,20+,21-,22-,23-/m0/s1 |
| Solution Type | Neat |
| SMILES | CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3C(=O)C[C@]12C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction