accessibility menu, dialog, popup

UserName

CAS RN 124431-80-7

CH3NHOONNNNNH2HNOOHOHOHHCl

CAS RN 124431-80-7

IUPAC Name: 3-{4-[2-({6-amino-9-[5-(ethylcarbamoyl)-3,4-dihydroxyoxolan-2-yl]-9H-purin-2-yl}amino)ethyl]phenyl}propanoic acid hydrochloride
Synonyms: 3-{4-[2-({6-amino-9-[5-(ethylcarbamoyl)-3,4-dihydroxyoxolan-2-yl]purin-2-yl}amino)ethyl]phenyl}propanoic acid hydrochloride
Molecular Weight (g/mol): 535.99
Molecular Formula: C23H30ClN7O6
InChi Key: QPHVMNOEKKJYJO-UHFFFAOYNA-N
SMILES: Cl.CCNC(=O)C1OC(C(O)C1O)N1C=NC2=C(N)N=C(NCCC3=CC=C(CCC(O)=O)C=C3)N=C12
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results